Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mirus Bio™ TransIT-PRO™ Transfection Reagent

Catalog No. MIR5750 Shop All Mirus Bio LLC Products
Change view
Click to view available options
Quantity:
1 mL
10 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MIR5750 10 mL
MIR5740 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MIR5750 Supplier Mirus Bio™ Supplier No. MIR 5750
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

TransIT-PRO™ Transfection Reagent decreases time to produce usable protein by maximizing target protein yields through transient transfection; high, reproducible protein yield in suspension CHO and 293 cells.

Reach critical stages faster. Decrease time to produce usable protein by maximizing target protein yields through transient transfection. The TransIT-PRO™ Transfection Reagent uses animal-origin-free components designed for high and reproducible protein yield in suspension CHO and 293 derived cells. Since it is compatible with varied media formulations, the same media can be used for both transient and stable expression. TransIT-PRO™ outperforms linear PEI in protein yield, while providing a cost-effective alternative to FreeStyle™ MAX.
  • Suitable for both transient and stable transfection
  • Compatible with multiple CHO media formulations
  • Reproducible protein expression with minimal optimization
  • Outperforms linear PEI in protein yield and provides a cost-effective alternative to FreeStyle™ MAX

Warranty and Services

1 Year

Specifications

Content And Storage Store at -20°C
Cell Type Mammalian
Format Tube
Promoter None (Promoterless)
Transfection Technique Chemical Transfection
For Use With (Application) Transfection
Product Type Transfection Reagent
Quantity 10 mL
Sterility Sterile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.