Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mirus Bio™ TransduceIT™ Transduction Reagent

Catalog No. MIR6620 Shop All Mirus Bio LLC Products
Change view
Click to view available options
Quantity:
1.0 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MIR6620 1.0 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MIR6620 Supplier Mirus Bio™ Supplier No. MIR 6620
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

TransduceIT™ Transduction Reagent is an aqueous solution of hexadimethrine bromide, a cationic polymer. Greatly enhances retroviral transduction and transgene expression in both dividing and quiescent mammalian cells.

  • Aqueous solution of hexadimethrine bromide, a cationic polymer
  • Provided as 10 mg/mL solution

Recommended Storage

Store at -20°C. Product guatanteed 6 months from date of purchase.

Specifications

Content And Storage Store at -20°C
Description TransduceIT™ Transduction Reagent
Cell Type 293-derived Cells
Format Tube
Promoter None (Promoterless)
Transfection Technique Chemical Transfection
Concentration 10 mg/mL
For Use With (Application) Transfection, Transduction, Gene expression, Lentivirus production
Product Type Transduction Reagent
Quantity 1.0 mL
Sterility Sterile
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.