Learn More
Description
- Each frozen aliquot contains 1mL of pure, titered culture of Yersinia enterocolitica
- Identification of the organism was confirmed by 16S sequencing
- Biosafety level 2
- Must be used within Biological Safety level 2 facility or cabinet
IMPORTANT: Contains live material. Customer must submit an executed ZeptoMetrix MTA before vendor can ship order.
Specifications
Specifications
| Certifications/Compliance | ISO, 13485 |
| Content And Storage | -65°C |
| Form | Solution |
| Media Type | Vial |
| Product Type | Microbiology Media |
| For Use With (Application) | Cross-reactivity studies, limit of detection studies, Nucleic Acid or molecular testing, other viral based assays |
| Quantity | 1 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
