Learn More
TARGETMOL CHEMICALS INC I3MT-3 5MG
Supplier: TARGETMOL CHEMICALS INC T88515MG
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. I3MT-3 (HMPSNE) (HMPSNE) is a potent, selective and cell membrane permeability inhibitor of 3-mercaptopyruvate sulfur transferase (3MST), which targets the persulfated cysteine residues located at the active site of 3MST. Purity 99.86%
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.