Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ i7500 Industrial Label Printer

Catalog No. 19029992 Shop All Brady Corporation Products
Change view
Click to view available options
Includes:
Software
Software, Autocutter
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
19-029-992 Software
19-029-993 Software, Autocutter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-029-992 Supplier Brady™ Supplier No. 177226
Add to Cart
Add to Cart

High-performance 3 in. core label printing the way you never thought possible.

Get 40-second part changeovers with no adjustments, no trial-and-error print tests and no wasted labels. LabelSense™ Technology allows the printer, Brady i75-Series materials and Brady software to communicate; automating and eliminating tedious adjustments and label-wasting tasks.

Specifications

Product Type Label Printer
Color Blue, Gray
Display Type Color LCD Touchscreen
For Use With (Equipment) THT Series Printers, 2HX Series Printers, 3PS Series Printers, Brady Workstation, Brady Labels
Material Plastic
Print Technology Direct Thermal, Thermal Transfer
Length (English) 20.9 in.
Width (English) 11.5 in
Includes Software
Resolution 300 dpi
Software Type Product and Wire ID Software
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.