Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ IARS Recombinant Protein

Catalog No. 89025905 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-025-905 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-025-905 Supplier Abnova™ Supplier No. H00003376Q0125
Add to Cart
Add to Cart

Specifications

For Use With (Application) ELISA, Western Blot, Antibody Production, Protein Array
pH Range 8
Quantity 25 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.