Learn More
Therapak™ IATA Shipping Boxes for Ambient Transport
Ideal kit packing boxes for reference labs, clinical trials and pathology kits
$241.00 - $336.50
Catalog Number | Mfr. No. | Dimensions (L x W x H) Exterior | Type | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Dimensions (L x W x H) Exterior | Type | Price | Quantity & Availability | ||||
22-130-024
|
Therapak™
23650 |
8 x 4.25 x 4 in. | Medium Ambient IATA Transport Box |
Case of 50 for $241.00
|
|
||||
22-130-049
|
Therapak™
23405 |
5.5 x 4.125 x5.75 in. | Upright Small Box |
Case of 50 for $336.50
|
|
||||
22-130-050
|
Therapak™
23675 |
6 x 4.25 x 4 in. | Small Ambient IATA Transport Box |
Case of 50 for $282.00
|
![]() |
||||
22-131-487
|
Therapak™
23358G |
12.25 x 9.25 x 2.25 in. | Tray Type IATA Shipping Box |
Case of 50 for $328.00
|
|
||||
22-131-488
|
Therapak™
23409G |
11.25 x 10.75 x 4.25 in. | Side-Load IATA Shipping Box | N/A | N/A | N/A | |||
Description
- Each box has a specific tab-lock mechanism to keep boxes closed during transport
- Ambient shipping boxes ideal for kit packaging and outbound shipping of diagnostic kits
- Known as two-way shipper boxes, all ambient shipper boxes can be used for return transport to the laboratory in addition to outbound shipping
Compliance
Meets IATA Packing Instruction 650 dimensional requirements.