Learn More
Abcam IB-MECA, A3 agonist, 5MG
Supplier:  Abcam AB1204385MG
Encompass_Preferred
MW 510.3 Da, Purity >97%. Highly potent and selective A3 agonist (Ki values are 1.1, 12.6 and 56 nM at A3, A1 and A2A receptors, respectively). Produces potent cardioprotective effects against myocardial injury. Mediates suppression of melanoma cells and suppresses human breast cancer cell proliferation.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7590
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.