Learn More
Description
Store serum up to seven days when it is essential to prevent any exchange of ions.
- One-way valve permits a physical gap to be created between the serum/plasma and clot, and prevents backflow into the primary collection tube
- Use with glass or plastic collection tubes
- Technique reduces second centrifugation, tube labeling and sample preparation time
- Allows direct sampling into automatic laboratory equipment
- Eliminates fibrin, particulate matter and equipment downtime
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Length (Metric) | 70mm |
| For Use With (Equipment) | Glass or Plastic collection tubes |
| For Use With (Application) | Eliminates fibrin, particulate matter and equipment downtime |
| Length (English) | 2.75 in. |
| Material | Polyethylene |
| Quantity | 100 Pack |
| Diameter (English) Outside | 0.5 in. |
| Diameter (Metric) Outside | 13mm |
| Product Type | Serum Filter |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
