Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ IB™ Model Serum Filters
GSA_VA

Catalog No. 0268166 Shop All Fisherbrand Products
Change view
Click to view available options
Length (English):
2.75 in.
4 in.
6 in.
Length (Metric):
70mm
100mm
152mm
Diameter (English) Outside:
0.5 in.
0.6 in.
Diameter (Metric) Outside:
13mm
16mm
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English) Length (Metric) Diameter (English) Outside Diameter (Metric) Outside
02-681-66 4 in. 100mm 0.5 in. 13mm
02-681-65 2.75 in. 70mm 0.5 in. 13mm
02-681-63 4 in. 100mm 0.6 in. 16mm
02-681-62 2.75 in. 70mm 0.6 in. 16mm
02-681-64 6 in. 152mm 0.6 in. 16mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 02-681-66
Add to Cart
Add to Cart
This item is not returnable. View return policy

Store serum up to seven days when it is essential to prevent any exchange of ions.

  • One-way valve permits a physical gap to be created between the serum/plasma and clot, and prevents backflow into the primary collection tube
  • Use with glass or plastic collection tubes
  • Technique reduces second centrifugation, tube labeling and sample preparation time
  • Allows direct sampling into automatic laboratory equipment
  • Eliminates fibrin, particulate matter and equipment downtime

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Diameter (English) Outside 0.5 in.
For Use With (Equipment) Glass or Plastic collection tubes
Quantity 100 Pack
Product Type Serum Filter
Length (English) 4 in.
For Use With (Application) Eliminates fibrin, particulate matter and equipment downtime
Material Polyethylene
Diameter (Metric) Outside 13mm
Length (Metric) 100mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.