Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IBI Scientific™ Blot Washer Trays

Catalog No. 15453257 Shop All Stovall Life Science Inc. Products
Change view
Click to view available options
Type:
Large Blot Tray
Mini Blot Tray
Standard Blot Tray
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
15-453-257 Large Blot Tray
15-453-252 Mini Blot Tray
15-453-253 Standard Blot Tray
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 15-453-257 Supplier IBI Scientific™ Supplier No. ACCBW0004
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For use with Blot Washer system, Belly Dancer Shaker and Hi/Lo Rocker

  • Comes with a lid
  • Available in MINI, Standard and Large

Specifications

Color Clear
For Use With (Equipment) Blot Washer System
Type Large Blot Tray
Material Polystyrene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.