Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ iBind™ Flex Cards

Catalog No. slf2010
Encompass_Preferred
Change view
Click to view available options
Type:
Card
Quantity:
10 Cards
40 Cards (10 x 4)
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Quantity
SLF2010 Card 10 Cards
SLF2010X4 Card 40 Cards (10 x 4)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. SLF2010 Supplier Invitrogen™ Supplier No. SLF2010
Only null left
Add to Cart
Edge
Add to Cart

Card for use with iBind™ Flex Western Blot Device.

  • Glass fiber backed card with filer paper stack at base
  • 10 cards per box
TRUSTED_SUSTAINABILITY

Specifications

Capacity 1 Midi Blot, 6 Vertically-Cut Strip Blots, 2 Mini Blots
Detection Method Chromogenic, Chemiluminescence, Fluorescence
For Use With (Application) Western Blot
For Use With (Equipment) iBind Flex Western Device
Membrane Compatibility Nitrocellulose, PVDF
Product Line iBind Flex
Running Time ∼2.5 hr.
Throughput 1 Midi Blot, 6 Vertically-Cut Strip Blots, 2 Mini Blots/Card
Type Card
Quantity 10 Cards
Shipping Condition Room Temperature
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.