Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ iBind PLUS Solution Kit

Catalog No. SLF1030
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 Kit
4 Kits
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SLF1030 1 Kit
SLF1030X4 4 Kits
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. SLF1030 Supplier Invitrogen™ Supplier No. SLF1030
Get 16% Off Protein Research Essentials
Use promo code PROTEIN16
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

The iBind PLUS Solution Kit increases signal sensitivity, requires less antibody per run, and simplifies western blotting immunoprocessing when used with the iBind Western System. The iBind PLUS Solution Kit comes with two ready to use solutions, iBind PLUS Solution and iBind PLUS Antibody Diluent.

  • The iBind PLUS Solution comes ready to use with no preparation required. It is used to prewet the iBind card prior to the run and acts as the block and wash buffer for the membrane processing, helping wash away unbound primary and secondary antibodies.
  • The iBind PLUS Antibody Diluent is used to dilute the primary and secondary antibodies. In most experiments, the primary antibody can be diluted to the same recommended concentration as used for traditional blotting, however only 2 mL of prepared primary antibody is required with iBind, saving up to 80% of primary antibody consumption. The iBind PLUS Antibody Diluent is specially formulated with bulking agents and stabilizing molecules to improve antibody binding with the target antigen to increase signal intensity.

During a run on the iBind, each solution is released from the iBind device wells through sequential lateral flow (SLF) and are wicked over the iBind card towards the stack region, incubating the membrane with each reagent. The glass fiber matrix of the iBind card allows for homogenous and consistent flow of the solutions to the membrane, increasing the antigen-antibody interaction.

The iBind PLUS Solution Kit, together with the iBind Western Device (Cat. No. SLF1000) and iBind Cards (Cat. No. SLF1010), bring automation and simplicity to western blotting with efficient and hands-free immunoprocessing in as little as 3 hours. Compared to manual methods, the iBind western system requires less primary antibody, enables reproducible results, and is compatible with downstream detection protocols.

Each iBind PLUS Solution Kit contains sufficient reagents for 10 mini-size blot immunoprocessing runs.

For iBind Flex, use the iBind Flex PLUS Solution Kit. For fluorescent applications, use the iBind Fluorescent Detection (FD) Solution Kit (Cat. No. SLF1019) or iBind Flex Fluorescent Detection (FD) Solution Kit (Cat. No. SLF2019).

TRUSTED_SUSTAINABILITY

Specifications

Membrane Compatibility Nitrocellulose, PVDF
Gel Compatibility Mini Precast Protein Gels
No. of Reactions 10 Mini Blots
Product Type Western Blot Kit
For Use With (Equipment) iBind Western Device
Content And Storage iBind PLUS Solution, iBind PLUS Antibody Diluent
Store at 2°C to 8°C
Kit Contents iBind PLUS Solution 250 mL, iBind PLUS Antibody Diluent, 45 mL
Detection Method Colorimetric, Chromogenic, Chemiluminescence
Product Line iBind
Quantity 1 Kit

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.