Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ iBind™ Western Starter Kit, fluorescent detection

Catalog No. SLF1002S
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SLF1002S 1 kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SLF1002S Supplier Invitrogen™ Supplier No. SLF1002S
  • $1,748.00
    $1,136.20 / Each of 1

    Save $611.80

    35% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The iBind Western Starter Kit for fluorescent detection includes an iBind Western Device, iBind Cards, and an iBind Fluorescent Detection Solution Kit, so you can get started with automated blot processing right away.

The iBind Western Device is a small benchtop device that automates western blot processing (immunodetection). It applies mechanical pressure to an iBind Card to generate sequential lateral flow (SLF) of immunodetection solutions for the blocking, antibody binding, and washingsteps. The glass fiber matrix of the iBind Card allows for homogenous and consistent flow of the solutions over the membrane, increasing the antigen-antibody interaction.

The iBind Western Device allows all solutions to be prepared and loaded in the device at the start of the procedure; all subsequent steps proceed automatically through SLF action—no electricity or batteries required. Blots are ready for the final detection steps in approximately 2.5 hours.

The iBind Western Device works exclusively with iBind Cards, and the iBind Solution Kit or iBind Fluorescent Detection (FD) Solution Kit to process one mini-blot.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 1 iBind Western Device, 10 iBind Cards (store at room temperature), 1 iBind Fluorescent Detection (FD) Solution Kit (store at 2 to 8°C)
Capacity 1 Mini Blot
Detection Method Fluorescence
For Use With (Application) Western Blot
Membrane Compatibility Nitrocellulose, PVDF
Product Line iBind
Running Time ∼2.5 hr.
Throughput 1 Mini Blot
Type Starter Kit
Quantity 1 kit
Shipping Condition Room Temperature
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.