Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ iBlot™ 2 Starter Kit

Catalog No. IB21001S
Encompass_Preferred
Change view
Click to view available options
Type:
Starter Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
IB21001S Starter Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. IB21001S Supplier Invitrogen™ Supplier No. IB21001S

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

View Alternate Product

The iBlot 2 Starter kit includes the iBlot 2 Gel Transfer Device as well as iBlot 2 Transfer Stacks (nitrocellulose) - everything needed to perform a rapid dry transfer of proteins from a gel to a membrane.

The iBlot 2 Starter Kit includes a iBlot 2 Gel Transfer Device that performs western blotting transfer simply, efficiently, and reliably, within seven minutes without the need for liquid buffers. The iBlot 2 system is compatible with both polyvinylidene difluoride (PVDF) and nitrocellulose membranes, and has comparable performance to traditional wet transfer methods in a fraction of the time. The iBlot 2 Gel Transfer Device is an integral part of the iBlot 2 Dry Blotting System, which consists of the transfer device and consumable transfer stacks that contain the required buffers and transfer membrane.

How the iBlot 2 System works
Buffer ion reservoirs are incorporated into the gel matrix (transfer stacks) instead of buffer tanks or soaked papers. The high density of ions in the gel matrix enables rapid protein transfer. During blotting, the copper anode does not generate oxygen gas as a result of water electrolysis, reducing blot distortion unlike conventional protein transfer techniques (including wet, semi-wet, and semi-dry) which use inert electrodes that generate oxygen. With the iBlot 2 Dry Blotting System, transfer time is reduced by the shortened distance between electrodes, high field strength, and high current. Trapped air bubbles, often created during the manual preparation of the blotting sandwich layers, are easy to avoid due to our unique de-bubbling design that promotes even and complete transfer.

  • Simple, user-friendly system
  • Complete protein transfer in 7 minutes or less
  • High detection sensitivity and even transfers
  • Increased blotting reliability and reproducibility
  • Flexible gel size formats and membrane types
  • Ability to create new custom programs
  • Built-in tutorial and application notes
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • iBlot 2 Gel Transfer Device
• iBlot 2 Transfer Stacks, nitrocellulose, regular size (2 boxes, 20 stacks)

For Use With (Application) Western Blot
Gel Compatibility Bis-Tris Gel, E-PAGE™ Gel, Midi, Mini, Tricine Gel, Tris-Acetate Gel, Tris-Glycine Gel
Membrane Compatibility Nitrocellulose, PVDF
Mode of Transfer Dry
Product Line iBlot 2
Running Time ∼7 min.
Throughput 1 Midi or 2 Mini Gels
Type Starter Kit
Quantity 1 Kit
Shipping Condition Approved for shipment at Room Temperature and wet ice
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.