Learn More
Invitrogen™ iBlot™ 3 Western Blot Transfer Device
Description
The iBlot 3 Western Blot Transfer Device is a dry transfer device that performs western blotting transfer efficiently and reliably in as few as three minutes and without the need for liquid buffers. The iBlot 3 system is compatible with both nitrocellulose and polyvinylidene difluoride (PVDF) membranes and offers equivalent or better performance compared with traditional wet-transfer methods in a fraction of the time. The iBlot 3 Western Blot Transfer Device is an integral part of the iBlot dry blotting system, which consists of the transfer device and consumable transfer stacks that contain the required buffers and transfer membrane (nitrocellulose or PVDF).
iBlot 3 Western Blot Transfer Device features include:
- Complete protein transfer in as few as three minutes
- High detection sensitivity and even, consistent protein transfer
- Two independently controlled transfer stations capable of transferring up to two midi or four mini blots
- Built-in adjustable cooling to ensure consistent transfer temperature
- Pre-programed transfer methods optimized for low, high, and broad molecular weight ranges
- Ability to create custom programs
- A simple, user-friendly interface
How the iBlot dry blotting system works
Buffer ion reservoirs are incorporated into the gel matrix of transfer stacks instead of buffer tanks or soaked filter papers. The high density of ions in the gel matrix enables rapid protein transfer. During blotting, the copper anode does not generate oxygen gas as a result of water electrolysis, reducing blot distortion (conventional protein transfer techniques, including wet, semi-wet, and semi-dry, use inert electrodes that generate oxygen).
With the iBlot dry blotting system, transfer time is reduced by the shortened distance between electrodes, high field strength, and high current. Trapped air bubbles, often created during the manual preparation of the blotting sandwich layers, are easy to avoid due to our unique de-bubbling design that promotes even and complete transfer. With the iBlot 3 system, there is no need to prepare buffers or clean up after blotting. The total preparation and run time is normally less than ten minutes per blot.
Specifications
Specifications
| Content And Storage | • iBlot 3 Gel Transfer Device • Western Blot Roller • REX Extended Warranty Store at room temperature. |
| For Use With (Application) | Western Blot |
| For Use With (Equipment) | iBlot 3 Transfer Stacks |
| Gel Compatibility | Mini- or Midi-sized: Bis-Tris Gels, Tricine Gels, Tris-Acetate Gels, and Tris-Glycine Gels |
| Membrane Compatibility | Nitrocellulose, PVDF |
| Mode of Transfer | Dry |
| Product Line | iBlot 3 |
| Running Time | ∼3 to 10 min. |
| Throughput | 2 Midi or 4 Mini Gels |
| Type | Transfer Device |
| Show More |
Frequently Asked Questions (FAQs)
Due to the thickness of the E-PAGE gels, they are not compatible with the iBlot3 device for transfer. Recommended methods for transfer of E-PAGE gels are semi-dry blotting or semi-wet blotting in the XCell II Blot Module. Methods are described in the E-PAGE Technical Guide.
No, we do not provide service contracts or extended warranties for these devices. The warranty is limited to 1 year from the date of shipment.
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.