Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IBMX (3-isobutyl-1-methylxanthine), MP Biomedicals™

Catalog No. MP218384891 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP218384891 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP218384891 Supplier MP Biomedicals, Inc Supplier No. 0218384891
Add to Cart
Add to Cart

A purine base found in most human body tissues and fluids and in other organisms. Several stimulants are derived from xanthine including caffeine, theophyline, and theobromine.

  • IBMX (3-isobutyl-1-methylxanthine), like other methylated xanthine derivatives, is a competitive non-selective phosphodiesterase inhibitor which raises intracellular cAMP, activates PKA, inhibits TNFα, and leukotriene synthesis
  • It reduces inflammation and innate immunity, and nonselective adenosine receptor antagonist
TRUSTED_SUSTAINABILITY

Specifications

Quantity 1 g
Color White
Purity >99%
Molecular Weight (g/mol) 222.3
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.