Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Ibotenic Acid, Excitotoxic Agonist, >98% Purity, 10 mM/mL

Catalog No. 502704534
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mM/mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-4534 10 mM/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-4534 Supplier Abcam Supplier No. AB14667010MM_ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Neuroexcitatory amino acid and potent NMDA and metabotropic receptor agonist used to model cognitive dysfunctions.

  • Molecular Weight: 158.11 Da
  • Purity: >98%
  • Description: Neuroexcitatory amino acid originally isolated from Amanita species. Neurotoxin often used to model cognitive dysfunctions.
  • Activity: NMDA and metabotropic receptor agonist.
  • Storage: Store at -20°C.
  • Shipping: Ambient - Can Ship with Ice.

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule NMDA Receptor, Metabotropic Receptor
Substrate Properties Chemical Substrate
Shipping Condition Ambient
Buffer 98% α-Amino-3-hydroxy-5-isoxazoleacetic acid
Form Solid
Content And Storage Store at -20°C.
Formulation 98% α-Amino-3-hydroxy-5-isoxazoleacetic acid
Quantity 10 mM/mL
For Use With (Application) Cognitive dysfunction modeling, NMDA receptor agonist, metabotropic receptor agonist
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.