Learn More
Advanced ImmunoChemical, Inc. ANTI-IGFBP-4 MAB
Supplier:  Advanced ImmunoChemical, Inc. 2IGFBP41MG
Encompass_Preferred
MONOCLONAL ANTIBODIES/MAbs IBP144, IBP154 and IBP180:human IGFBP-4; human N-terminal IGFBP-4 fragment (NT-IGFBP-4)MAbs IBP182, IBP185 and IBP190:human IGFBP-4; human C-terminal IGFBP-4 fragment (CT-IGFBP-4)MAb IBP3:human NT-IGFBP-4, cross-reaction with full-length human IGFBP-4MAb IBP163:human CT-IGFBP-4, cross-reaction with full-length human IGFBP-4 <5% (ELISA)Detection of human IGFBP-4, NT-IGFBP-4, and CT-IGFBP-4. MAbs are working in direct ELISA and sandwich immunoassay.Recommended pairs for human IGFBP-4 sandwich immunoassay (capture-detection): IBP185 –IBP154 IBP182 – IBP144 IBP144 – IBP190 Recommended pairs for specific human NT-IGFBP-4 sandwich immunoassay (capture-detection):IBP3 – IBP144 IBP3 – IBP180 Recommended pairs for specific human CT-IGFBP-4 sandwich immunoassay (capture-detection): IBP182 – IBP163Purification:Chromatography on protein A SepharosePresentation:PBS, pH 7.4, 0.1 % sodium azide (NaN3)Storage Conditions:+ 4 °C
Catalog No. NC3455104