Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam IBTP, Mitochondrial Thiol Alkylation Reagent, Solid, 25 mg

Catalog No. 502705532
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-5532 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-5532 Supplier Abcam Supplier No. AB14464625MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cell-permeable reagent for specific labelling of mitochondrial thiols to identify thiol redox state.

  • Molecular weight: 572.2 Da
  • Purity: >98%
  • Physical state: Solid
  • Function: Mitochondrial thiol-specific cationic alkylation reagent
  • Applications: Selective mitochondrial labeling and thiol redox state identification
  • Cell permeability: Yes
  • CAS number: 159085-21-9
  • Composition: 100% 4-iodobutyl(triphenyl)phosphanium;iodide

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule Thiols (Mitochondrial)
Substrate Properties Chemical Substrate
Shipping Condition Ambient
Buffer 100% 4-iodobutyl(triphenyl)phosphanium iodide
Content And Storage Store at -20°C.
Form Solid
Formulation 100% 4-iodobutyl(triphenyl)phosphanium iodide
Quantity 25 mg
For Use With (Application) Labelling of mitochondrial thiols, identify thiol redox state
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.