Learn More
Abcam IBTP, Mitochondrial Thiol Specific Alkylation Reagent, 5 mg
Description
- Molecular Weight: 572.2 Da
- Purity: >98%
- Form: Solid
- Properties: Mitochondrial thiol-specific cationic alkylation reagent; selective uptake into mitochondria leads to specific labelling of mitochondrial thiols; can be used to identify thiol redox state
- Cell-Permeable: Yes
- Storage Buffer: Constituents: 100% 4-iodobutyl(triphenyl)phosphanium; iodide
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | Thiols |
| Substrate Properties | Chemical Substrate |
| Shipping Condition | Ambient |
| Buffer | 100% 4-iodobutyl(triphenyl)phosphanium;iodide |
| Form | Solid |
| Content And Storage | Store at -20°C. |
| Formulation | 100% 4-iodobutyl(triphenyl)phosphanium;iodide |
| Quantity | 5 mg |
| For Use With (Application) | Mitochondrial thiol labelling, thiol redox state identification |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.