Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ibudilast, Thermo Scientific™

Catalog No. AC469782500
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC469782500 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AC469782500 Supplier Thermo Scientific Chemicals Supplier No. 469782500
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Chemical Name or Material Ibudilast
CAS 50847-11-5
Synonym 3-Isobutyryl-2-isopropylpyrazolo[1,5-a]pyridine
Quantity 250 mg
Melting Point 54°C to 58°C
Shipping Condition No special requirements
Product Line Life science
Molecular Formula C14 H18 N2O
Physical Form Crystalline Powder
Color White to Almost-white
Purity ≥97.5%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.