Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ibuprofen, USP, 97-103%, Spectrum™ Chemical
SDP

Catalog No. 18603405 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
5 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-603-405 100 g Poly Bottle
18-603-408 5 kg Poly Pail
18-603-409 50 kg Fiber Drum
18-603-404 25 g Poly Bottle
18-603-406 500 g Poly Bottle
18-603-407 1 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18-603-405 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. IB100100GM
Only null left
Add to Cart
Add to Cart

IB100, 15687-27-1, C13H18O2

Spectrum™ Chemical Ibuprofen, USP is a non-steroidal anti-inflammatory drug often used to treat and reduce fevers and swelling. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 15687-27-1
Molecular Formula C13H18O2
Molecular Weight (g/mol) 206.29
MDL Number MFCD00010393
InChI Key HEFNNWSXXWATRW-UHFFFAOYNA-N
IUPAC Name 2-[4-(2-methylpropyl)phenyl]propanoic acid
SMILES CC(C)CC1=CC=C(C=C1)C(C)C(O)=O

Specifications

CAS Max % 100%
Ignition Residue 0.5%
Packaging Poly Bottle
Quantity 100 g
Percent Purity 97 to 103%
Grade USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.