Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ibuprofen, USP grade, MP Biomedicals™

Catalog No. 0219021605 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
02-190-21605 5 g
ICN19021601 1 g
ICN19021610 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 02-190-21605 Supplier MP Biomedicals, Inc Supplier No. 0219021605
Add to Cart
Add to Cart

Ibuprofen is clinically useful as a nonsteroidal anti-inflammatory drug.

  • Cyclooxygenase (COX) inhibitor that has greater activity against COX-1 than against COX-2.
  • Also inhibits cyclooxygenase, PGH synthase-1 and PGH synthase-2.
  • Ibuprofen is reported to inhibit the netrophil-dependent edema response of rabbit skin to C5a/PGE23.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15687-27-1
Molecular Formula C13H18O2
Molecular Weight (g/mol) 206.29
MDL Number MFCD00010393
InChI Key HEFNNWSXXWATRW-UHFFFAOYNA-N
Synonym ibuprofen, 2-4-isobutylphenyl propanoic acid, motrin, brufen, nurofen, advil, dolgit, liptan, nuprin, anflagen
PubChem CID 3672
ChEBI CHEBI:5855
IUPAC Name 2-[4-(2-methylpropyl)phenyl]propanoic acid
SMILES CC(C)CC1=CC=C(C=C1)C(C)C(O)=O

Specifications

Melting Point 75 to 77.5°C
Color White
Quantity 5 g
Solubility Information Water Solubility: 21 mg/L (at 25°C)
Vapor Pressure 4.74X10-5 mm Hg at 25°C
Formula Weight 206.3
Grade USP
Physical Form Powder
Chemical Name or Material Ibuprofen
EINECSNumber 239-784-6
Recommended Storage Store at Room Temperature (15 to 30°C)

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.