Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Ibutilide Fumarate, Sodium Channel Activator, Solid, 20 mM/mL

Catalog No. 502704505
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 mM/mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-4505 20 mM/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-4505 Supplier Abcam Supplier No. AB14673820MM_ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sodium channel activator that increases the refractory period and prolongs myocardial action potential duration.

  • Purity: >99%
  • Molecular Weight: 500.7 Da
  • CAS Number: 122647-32-9
  • Form: Solid
  • Activity: Sodium channel activator; activates inward depolarising slow sodium currents, increases the refractory period, and prolongs myocardial action potential duration. Class III antiarrhythmic.
  • Storage: Store at Ambient.
  • Shipping: Ambient - Can Ship with Ice
  • Composition: 100% Ibutilide Hemifumarate

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule Sodium Channel
Form Solid
Shipping Condition Ambient
Concentration 20 mM
Content And Storage Store at ambient temperature.
Buffer 100% Ibutilide Hemifumarate
Formulation 100% Ibutilide Hemifumarate
Quantity 20 mM/mL
For Use With (Application) Sodium channel activation
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.