Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CritiCore Protective Wear IC03 Non-Linting Cleanroom Frock

Catalog No. 17604254
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
6X-Large
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-604-254 2X-Large
17-604-250 Small
17-604-251 Medium
17-604-252 Large
17-604-253 X-Large
17-604-255 3X-Large
17-604-256 4X-Large
17-604-257 5X-Large
17-604-258 6X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Catalog No. 17-604-254 Supplier CritiCore Protective Wear Supplier No. 12F23165IC03WK0000WHT2XL
Add to Cart
Add to Cart
This item is not returnable. View return policy

This IC03 Non-Linting Cleanroom Frock is designed to restrict particulates in controlled environments, and provides excellent ESD properties. This fabric has been engineered with a filament polyester rated for ISO class 5 cleanrooms and above.

  • IC03 Fabric
  • Grid-ESD / Non-Linting
  • Knit-Cuff Sleeves
  • Coil Zipper Front Enclosure
  • Neck Snaps at top of Mandarin Collar
  • Primary Application: Microelectronics, Semi-Conductor, Precision Optics, Wafer Manufacturing

Specifications

Size 2X-Large
Material Polyester
Color White
Disposable Reusable
Sterility Non-sterile
Closure Type Zipper
Product Type Cleanroom Frock
Seam Type Stitched
For Use With (Application) Restrict particulates in controlled environments
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.