Learn More
CritiCore Protective Wear IC03 Pullover Cleanroom Hood
Description
- IC03 Fabric
- Grid-ESD / Non-Linting
- Pullover Hood
- Snap and Elastic Adjustment
- Primary Applications: Microelectronics, Semi-Conductor, Precision Optics, Wafer
Specifications
Specifications
| Product Type | Cleanroom Hood |
| Size | Small |
| Material | Polyester |
| Color | White |
| Disposable | Reusable |
| Cleanroom Class | Class 100 (ISO 5) |