Learn More
CritiCore Protective Wear IC03 Pullover Cleanroom Hood
Catalog No. 17604247
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-604-247 Large
17-604-244 X-Small
17-604-245 Small
17-604-246 Medium
17-604-248 X-Large
17-604-249 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 17-604-247 Supplier CritiCore Protective Wear Supplier No. 11HD2470IC03WK0000WHTL
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
This IC03 non-linting cleanroom hood is designed to restrict particulates in controlled environments, and provides excellent ESD properties. This fabric has been engineered with a filament polyester rated for ISO class 5 cleanrooms and above.
  • IC03 Fabric
  • Grid-ESD / Non-Linting
  • Pullover Hood
  • Snap and Elastic Adjustment
  • Primary Applications: Microelectronics, Semi-Conductor, Precision Optics, Wafer
Specifications
Product Type Cleanroom Hood
Size Large
Material Polyester
Color White
Disposable Reusable
Cleanroom Class Class 100 (ISO 5)