Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ICA1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA544208
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA544208 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA544208 Supplier Invitrogen™ Supplier No. PA544208
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: SKAIVLYQKR ICFLSQEENE LGKFLRSQGF QDKTRAGKMM QATGKALCFS Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 86%.

This gene encodes a protein with an arfaptin homology domain that is found both in the cytosol and as membrane-bound form on the Golgi complex and immature secretory granules. This protein is believed to be an autoantigen in insulin-dependent diabetes mellitus and primary Sjogren's syndrome. Alternatively spliced variants which encode different protein isoforms have been described; however, not all variants have been fully characterized.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ICA1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ICA1
Gene Accession No. Q05084
Gene Alias 69 kDa islet cell autoantigen; 69kDa; diabetes mellitus type I autoantigen; ica1; ica1 {ECO:0000312; ICA69; ICAp69; Islet cell autoantigen 1; islet cell autoantigen 1 isoform; islet cell autoantigen 1, 69 kDa; islet cell autoantigen 1, 69kDa; islet cell autoantigen p69; p69; RGD:621465}; testicular tissue protein Li 162; Unknown (protein for MGC:134001); zgc:92566
Gene Symbols ICA1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human ICA1 (aa 74-123).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3382
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.