Learn More
Invitrogen™ ICAM-1 Monoclonal Antibody (D3.6)

Description
ICAM-1 Monoclonal Antibody for Flow
Specifications
Specifications
| Antigen | ICAM-1 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | D3.6 |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.08% sodium azide |
| Gene | ICAM1 |
| Gene Accession No. | P05362 |
| Gene Alias | BB2; CD54; cell surface glycoprotein P3.58; Human rhinovirus receptor; ICAM; Icam1; ICAM-1; ICAM-1; CD54 homolog; Intercellular adhesion molecule 1; intercellular adhesion molecule 1 (CD54), human rhinovirus receptor; intercellular adhesion molecule-1; intercellular adhesion molecule-1 precursor; Ly 47; Ly-47; major group rhinovirus receptor; MALA2; MALA-2; MyD10; P3.58; sCD54; soluble CD54; Surface antigen of activated B cells |
| Gene Symbols | ICAM1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.