Learn More
ICAM-1 Mouse anti-Human, Rat, FITC, Clone: 2F9A8, Proteintech
Shop All Proteintech Group Inc Products

Description
This antibody is FITC conjugated ICAM1 mouse monoclonal antibody (600299-1-Ig) which has passed in WB, IHC and FC detection.
Specifications
Specifications
| Antigen | ICAM-1 |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 2F9A8 |
| Concentration | 0.27 mg/mL |
| Conjugate | FITC |
| Formulation | PBS with 50% glycerol and 0.05% Proclin 300; pH 7.3 |
| Gene | ICAM1 |
| Gene Accession No. | P05362, Q00238 |
| Gene Alias | CD54, ICAM1, ICAM-1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.