Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ICAM-2 Monoclonal Antibody (3C4), Alexa Fluor™ 647
GREENER_CHOICE

Catalog No. 501137057
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-113-7057 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-113-7057 Supplier Invitrogen™ Supplier No. A15452
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rat Monoclonal Antibody

ICAM-2 Monoclonal Antibody for Flow

ICAM-2 is a member of the intercellular adhesion molecule (ICAM) family. All ICAM proteins are type I transmembrane glycoproteins, contain 2-9 immunoglobulin-like C2-type domains, and bind to the leukocyte adhesion LFA-1 protein. ICAM-2 may play a role in lymphocyte recirculation by blocking LFA-1-dependent cell adhesion. It mediates adhesive interactions important for antigen-specific immune response, NK-cell mediated clearance, lymphocyte recirculation, and other cellular interactions important for immune response and surveillance. Several transcript variants encoding the same protein have been found for this gene. Diseases associated with ICAM2 include Colon Carcinoma In Situ.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ICAM-2
Applications Flow Cytometry
Classification Monoclonal
Clone 3C4
Concentration 0.5 mg/mL
Conjugate Alexa Fluor 647
Formulation PBS with <0.1% sodium azide; pH 7.4
Gene ICAM2
Gene Accession No. P35330
Gene Alias CD102; Icam2; ICAM-2; Intercellular adhesion molecule 2; Ly-60; Lymphocyte function-associated AG-1 counter-receptor; RP23-418O11.10-001
Gene Symbols ICAM2
Host Species Rat
Immunogen Mouse CD102 from COS cells transfected with mouse ICAM-2 cDNA and BW5147 AKR/J mouse thymoma cell line.
Purification Method Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15896
Target Species Mouse
Content And Storage 4°C, store in dark
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.