Learn More
Invitrogen™ ICAM-3 Monoclonal Antibody (MEM-04)

Description
This antibody reacts with an extracellular epitope of CD50 (ICAM-3), a 120-130 kDa type I membrane protein (immunoglobulin supergene family) expressed on leukocytes, endothelial cells and Langerhans cells; it is negative on platelets and erythrocytes. The MEM-04 blocks DC-SIGN (CD209) binding to ICAM-3 (epitope in D1). Functional Application: Blocking.
Specifications
Specifications
| Antigen | ICAM-3 |
| Applications | Flow Cytometry, Functional Assay, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | MEM-04 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | ICAM3 |
| Gene Accession No. | P32942 |
| Gene Alias | CD50; CDw50; ICAM3; ICAM-3; ICAM-R; Intercellular adhesion molecule 3; intercellular adhesion molecule-3 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.