Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Icarapin Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144566
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144566 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144566 Supplier Invitrogen™ Supplier No. PA5144566
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Icarapin Polyclonal Antibody for Western Blot, ELISA

Icarapin is a peptide that is derived from the venom of the South American pit viper, Bothrops jararaca. It has been studied for its potential therapeutic properties, particularly its antimicrobial and antifungal activities. Icarapin has shown inhibitory effects against a range of bacteria and fungi, including drug-resistant strains. Research is ongoing to further explore the potential applications and mechanisms of action of icarapin, as well as its potential as a source for the development of new antimicrobial agents.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Icarapin
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene Alias Icarapin
Host Species Rabbit
Immunogen Recombinant Apis mellifera carnica Icarapin protein (20-223aa).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Insect
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.