Learn More
Abbott™ iCassette™ Urine Drug Screen
Catalog No. 22911018
Change view
Click to view available options
Detectable Analytes:
Cocaine (COC), Cannibinoids (THC), Opiates (OPI), Amphetamine (AMP), Methamphetamine (mAMP), Phencyclidine (PCP), Benzodiazepines (BZO), Barbiturates (BAR)
Cocaine (COC), Cannibinoids (THC), Opiates (OPI), Amphetamine (AMP), Methamphetamine (mAMP), Phencyclidine (PCP), Benzodiazepines (BZO), Barbiturates (BAR), Methadone (MTD)
Cocaine (COC), Cannibinoids (THC), Opiates (OPI), Amphetamine (AMP), Methamphetamine (mAMP), Phencyclidine (PCP), Benzodiazepines (BZO), Barbiturates (BAR), Methadone (MTD), Oxycodone (OXY)
Cocaine (COC), Cannibinoids (THC), Opiates (OPI), Amphetamine (AMP), Methamphetamine (mAMP), Phencyclidine (PCP), Benzodiazepines (BZO), Barbiturates (BAR), Methadone (MTD), Tricyclic Antidepressants (TCA)
Cocaine (COC), Cannibinoids (THC), Opiates (OPI), Methamphetamine (mAMP)
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Detectable Analytes
22-911-018 Cocaine (COC), Cannibinoids (THC), Opiates (OPI), Methamphetamine (mAMP)
22-911-024 Cocaine (COC), Cannibinoids (THC), Opiates (OPI), Amphetamine (AMP), Methamphetamine (mAMP), Phencyclidine (PCP), Benzodiazepines (BZO), Barbiturates (BAR), Methadone (MTD)
22-911-023 Cocaine (COC), Cannibinoids (THC), Opiates (OPI), Amphetamine (AMP), Methamphetamine (mAMP), Phencyclidine (PCP), Benzodiazepines (BZO), Barbiturates (BAR)
22-911-026 Cocaine (COC), Cannibinoids (THC), Opiates (OPI), Amphetamine (AMP), Methamphetamine (mAMP), Phencyclidine (PCP), Benzodiazepines (BZO), Barbiturates (BAR), Methadone (MTD), Oxycodone (OXY)
22-911-025 Cocaine (COC), Cannibinoids (THC), Opiates (OPI), Amphetamine (AMP), Methamphetamine (mAMP), Phencyclidine (PCP), Benzodiazepines (BZO), Barbiturates (BAR), Methadone (MTD), Tricyclic Antidepressants (TCA)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 22-911-018 Supplier Abbott™ Supplier No. IDOA1145
Add to Cart
Add to Cart
This item is not returnable. View return policy
One step screening test for the simultaneous detection of multiple drugs and metabolites in urine
Rapid urine screening test that can be performed without the use of an instrument
  • Simple procedure: collect and pipette to test — results in as little as 5 minutes
  • Requires only 5mL of specimen
  • Tests for up to 10 drugs at once
  • Results can be photocopied for record keeping purposes
  • 12-18 Month shelf life
  • Results are easy to read
  • Room temperature storage
Specifications
Detectable Analytes Cocaine (COC), Cannibinoids (THC), Opiates (OPI), Methamphetamine (mAMP)
Sample Type Urine
Type iCassette™ Urine Drug Screen
Quantity 25 Pack
Test Time 5 min.