Learn More
Abbott™ iCassette™ Urine Drug Screen
Description
Rapid urine screening test that can be performed without the use of an instrument
- Simple procedure: collect and pipette to test — results in as little as 5 minutes
- Requires only 5mL of specimen
- Tests for up to 10 drugs at once
- Results can be photocopied for record keeping purposes
- 12-18 Month shelf life
- Results are easy to read
- Room temperature storage
Specifications
Specifications
| Detectable Analytes | Cocaine (COC), Cannibinoids (THC), Opiates (OPI), Amphetamine (AMP), Methamphetamine (mAMP), Phencyclidine (PCP), Benzodiazepines (BZO), Barbiturates (BAR) |
| Sample Type | Urine |
| Type | iCassette™ Urine Drug Screen |
| Quantity | 25 Pack |
| Test Time | 5 min. |