Learn More
Cosmo Bio Usa Inc Ice On Type 2
SDP
Supplier:  Cosmo Bio Usa Inc SKBIO2
Encompass
Tubes will not be in direct contact with water and ice. Racks are made of aluminum, and are rust-free. Compact design. Can be stacked in the refrigerator or freezer. Can contain 96 tubes of 0.2 mL or 12 x 0.2 mL 8-tube strips or 8 x 0.2 mL 12-tube strips or 1 x 0.2 mL 96 well plate and 6 tubes of 0.5 mL and 4 tubes of 1.5 mL. Easy to be separated from tubes of 1.5 mL or of 0.5 mL to tubes of 0.2 mL.
Catalog No. NC1663277
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.