Learn More
Thomas Scientific Ice Pan, 4L, Polystyrene, Blue
SDP
Supplier:  Thomas Scientific 1189K78
Encompass
Lightweight yet durable high density expanded polystyrene ice buckets and pans. Our containers are produced with thick walls to provide rigidity and strength, and yet are super lightweight. These new Polsarsafe™ storage products are perfectly suited for use with ice, dry ice, slurry ice, etc.  The 3 pans and 2 buckets provide excellent insulating properties to help keep temperature sensitive samples safely chilled, but will not leave condensation on the benchtop. We purposely designed our containers for stable and secure benchtop use, but also added features to make them easy to carry from lab to lab. Our buckets come with snug  tting lids to minimize evaporation loss and to maximize cooling.  The Polarsafe™ buckets and pans are cool containers that are a less costly alternative to expensive and cumbersome polyurethane models. Each ice bucket comes with prominent recessed handles for easy transport, as well as a snug fitting lid which features easy to grab kn
Catalog No. 50-145-7347
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.