Learn More
Cellular Dynamics International iCell Endothelial Cells, 01434, 1 Vial: ≥ 1 x 106 cells/vial
Supplier:  Cellular Dynamics International R1022
Encompass
Endothelial cells are highly dynamic cells that form the inner lining of blood vessels. In addition to their structural role, endothelial cells regulate the passage of substances between the bloodstream and surrounding tissue. They also secrete mediators that influence vascular hemodynamics, inflammation, cell trafficking, and the remodeling and formation of new blood vessels. Endothelial cell dysfunction is associated with a variety of diseases including atherosclerosis, coronary artery disease, hypertension, and inflammatory disorders.
Catalog No. NC3449478
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.