Learn More
FUJIFILM BIOSCIENCES INC iCell Microglia Kit, 01279
Supplier:  FUJIFILM BIOSCIENCES INC R1131
Encompass
Each kit contains: C1110 Cryopreserved iCell Microglia, 01279, M1034 50ml iCell Glial Base Medium, M1036 0.5ml iCell Microglia Supplement A (100x), M1037 0.5ml iCell Microglia Supplement B (100x), M1035 2 x 1ml iCell Neural Supplement C (25X), X1022 iCell Microglia Quick Guide. ≥1 x 106 Viable Cells/Vial
Catalog No. NC1599538