Learn More
FUJIFILM BIOSCIENCES INC iCell Microglia Rett Syndrome MECP2, 01279, Isogenic Control, 1 Vial: > 1 x 106 cells/vial
Supplier:  FUJIFILM BIOSCIENCES INC C1110
Encompass
The engineered iCell Microglia, MECP2 line contains an insert directly after Serine 49 that includes stop codons in all 3 reading frames, generating a non-functional truncated protein before the methyl-CpG-binding domain resulting in the protein being p.(Ala50Ter). The MECP2 line was engineered in the otherwise healthy background line, 01279.
Catalog No. NC1824700