Learn More
Selleck Chemical LLC ICG-001 10mM/1mL 780757-88-2 (relative stereochemistry); 847591-62-2 (absolute stereochemistry)
SDP
Supplier:  Selleck Chemical LLC S266210MM/1ML
Encompass_Preferred
ICG-001 antagonizes Wnt/?-catenin/TCF-mediated transcription and specifically binds to CREB-binding protein (CBP) with IC50 of 3 ?M, but is not the related transcriptional coactivator p300. ICG-001 induces apoptosis. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-2703
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.