Description
Module consists of:
- VIS cassette
- UVA cassette
- Independent power supply
Specifications
Specifications
| For Use With (Equipment) | KB PRO/KBF/KBF PRO 260/470 |
| Material | Glass |
| Type | Lighting |