Description
Module consists of:
- VIS cassette
- UVA cassette
- Spherical VIA sensor
- Spherical UVA sensor
- Independent power supply
Specifications
Specifications
| For Use With (Equipment) | KBF/KBF PRO 260/470, KB PRO 260/470 |
| Material | Glass |
| Type | Lighting |
| Includes | LQC |