Learn More
Bioworld Ichihashi New Phalaenopsis (NP) Medium, 25 L
Supplier:  Bioworld 306301962
Encompass_Preferred
Ichihashi New Phalaenopsis (NP) MediumContains the Macronutrients and Micronutrients as described by Ichihashi (1992) Modified to contain NH4NO3Sucrose, Cytokines, Vitamins, and Auxins should be added as needed Provided as 1 unit to make 25L or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture testedWorking Concentration: 25.35 g/LSoluble in water
Catalog No. 50-254-989
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.