Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ichthammol, USP, Spectrum™ Chemical
SDP

Catalog No. 18603411 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Amber Glass Bottle
Drum
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-603-411 500 g Amber Glass Bottle
18-603-410 125 g Amber Glass Bottle
18-603-412 2.5 kg Amber Glass Bottle
18-603-413 12 kg Poly Pail
18-603-414 50 kg Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-603-411 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. IC110500GM
Add to Cart
Add to Cart

IC110, 8029-68-3

Spectrum™ Chemical Ichthammol, USP , also known as ammonium bituminosulfonate, is used to treat certain skin diseases like psoriasis and eczema. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 8029-68-3

Specifications

CAS Max % 100%
Ignition Residue 0.5%
Loss on Drying 50%
Packaging Amber Glass Bottle
Quantity 500 g
Grade USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.