Learn More
Selleck Chemical LLC ICI-118551 Hydrochloride 5mg 1217094-53-5 ICI-118,551
SDP
Supplier:  Selleck Chemical LLC S81145MG
Encompass_Preferred
ICI-118551 is a highly selective ?2-adrenergic receptor antagonist with Ki values of 0.7, 49.5 and 611 nM for ?2, ?1 and ?3 receptors, respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5671
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.