Learn More
Abcam ICI 118551 hydrochloride, beta2 adrenoceptor antagonist, 50MG
Supplier:  Abcam AB12080850MG
Encompass_Preferred
MW 313.9 Da, Purity >99%. Highly selective β2 adrenoceptor antagonist (Ki values are 1.2, 120 and 257 nM at β2, β1 and β3 receptors, respectively). Blood-brain barrier permeable. Decreases vascular tone *in vivo*.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7243