Learn More
Medchemexpress LLC Iclaprim | 192314-93-5 | MFCD09837803 | 98.9% | 354.40 g/mol | C19H22N4O3 | 100 MG
Supplier: Medchemexpress LLC HY101479100MG
Iclaprim is a bacterial dihydrofolate reductase inhibitor supplied as a research-grade powder for analytical and laboratory use. It demonstrates potent activity against Gram-positive bacteria, including methicillin-resistant Staphylococcus aureus, with a reported MIC90 of 0.06 μg/mL. Molecular formula C19H22N4O3; molecular weight 354.40 g/mol.
- Research-grade powder form for laboratory assays.
- High stated purity of 98.9% for reliable analytical results.
- Potent activity against Staphylococcus aureus (MRSA).
- Suitable as a reference standard or tool compound in microbiology studies.
- Available in 100 mg quantity for small-scale experiments.