Learn More
Medchemexpress LLC Iclaprim | 192314-93-5 | MFCD09837803 | >98.0% | 354.40 g·mol⁻¹ | C19H22N4O3 | 10 MG
Supplier: Medchemexpress LLC HY10147910MG
Iclaprim is a diaminopyrimidine antibiotic that inhibits bacterial dihydrofolate reductase and is active against Gram-positive organisms, including methicillin-resistant Staphylococcus aureus (MRSA). Supplied as a 10 mg research reagent, it is intended for in vitro studies and preclinical research. Molecular formula C19H22N4O3; molecular weight 354.40 g·mol⁻¹.
- Potent inhibitor of bacterial dihydrofolate reductase.
- Active against MRSA with reported MIC90 of 0.06 μg/mL.
- Provided as a small-scale research sample for in vitro assays.
- Well characterized chemically with defined formula and molecular weight.
- Purity commonly reported as >98% by HPLC in supplier listings.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.